SY3H3D682CD7 Synthonix Corporation

3-Oxoisoindoline-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 935269-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51213.html

Safety Information

MSDS https://synthonix.com/51213.html
Availability details for
Quantity
  • Availability
  • Add to Cart