SY3H3D682CD8 Synthonix Corporation

3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 483282-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/51227.html

Safety Information

MSDS https://synthonix.com/51227.html
Availability details for
Quantity
  • Availability
  • Add to Cart