SY3H3D682CDB Synthonix Corporation

3-Oxo-2-(pyridin-2-yl)butanenitrile

  • MDL number No MDL found
  • Cas Number : 57115-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51359.html

Safety Information

MSDS https://synthonix.com/51359.html
Availability details for
Quantity
  • Availability
  • Add to Cart