SY3H3D682CDD Synthonix Corporation

N-(2-Oxo-2-phenylethyl)formamide

  • Cas Number : 73286-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51780.html

Safety Information

MSDS https://synthonix.com/51780.html
Availability details for
Quantity
  • Availability
  • Add to Cart