SY3H3D682CDE Synthonix Corporation

(R)-Oxiran-2-ylmethyl 3-nitrobenzenesulfonate

  • MDL number No MDL found
  • Cas Number : 115314-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/51866.html

Safety Information

MSDS https://synthonix.com/51866.html
Availability details for
Quantity
  • Availability
  • Add to Cart