SY3H3D682CDF Synthonix Corporation

1-Oxa-9-azaspiro[5.5]undecane hydrochloride

  • MDL number No MDL found
  • Cas Number : 1414958-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/52200.html

Safety Information

MSDS https://synthonix.com/52200.html
Availability details for
Quantity
  • Availability
  • Add to Cart