SY3H3D682CEA Synthonix Corporation

3-(5-Oxazolyl)benzoic Acid

  • Cas Number : 252928-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53575.html

Safety Information

MSDS https://synthonix.com/53575.html
Availability details for
Quantity
  • Availability
  • Add to Cart