SY3H3D682CEC Synthonix Corporation

2-Oxo-1,2,3,4-tetrahydroquinoline-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 88371-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53968.html

Safety Information

MSDS https://synthonix.com/53968.html
Availability details for
Quantity
  • Availability
  • Add to Cart