SY3H3D682D22 Synthonix Corporation

2-(4-Oxo-4,5,6,7-tetrahydro-3H-cyclopenta[4,5]thieno[2,3-d]pyrimidin-3-yl)acetic acid

  • MDL number No MDL found
  • Cas Number : 371206-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54299.html

Safety Information

MSDS https://synthonix.com/54299.html
Availability details for
Quantity
  • Availability
  • Add to Cart