SY3H3D682D25 Synthonix Corporation

2-(4-(2-Oxoimidazolidin-1-yl)phenyl)acetic acid

  • MDL number No MDL found
  • Cas Number : 492445-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54493.html

Safety Information

MSDS https://synthonix.com/54493.html
Availability details for
Quantity
  • Availability
  • Add to Cart