SY3H3D682D2F Synthonix Corporation

N-(5-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)acetamide

  • MDL number No MDL found
  • Cas Number : 88611-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/55823.html

Safety Information

MSDS https://synthonix.com/55823.html
Availability details for
Quantity
  • Availability
  • Add to Cart