SY3H3D682D33 Synthonix Corporation

2-(4-oxo-3-Phenyl-1,3-selenazolidin-2-ylidene)malononitrile

  • MDL number No MDL found
  • Cas Number : 887000-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56396.html

Safety Information

MSDS https://synthonix.com/56396.html
Availability details for
Quantity
  • Availability
  • Add to Cart