SY3H3D682D34 Synthonix Corporation

2,2'-Oxybis(nitrobenzene)

  • MDL number No MDL found
  • Cas Number : 2217-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/56423.html

Safety Information

MSDS https://synthonix.com/56423.html
Availability details for
Quantity
  • Availability
  • Add to Cart