SY3H3D682D37 Synthonix Corporation

2-Oxo-5-phenyl-1,2-dihydropyridine-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 10177-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56524.html

Safety Information

MSDS https://synthonix.com/56524.html
Availability details for
Quantity
  • Availability
  • Add to Cart