SY3H3D682D38 Synthonix Corporation

4-(4-Oxocyclohexyl)benzonitrile

  • Cas Number : 73204-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56813.html

Safety Information

MSDS https://synthonix.com/56813.html
Availability details for
Quantity
  • Availability
  • Add to Cart