SY3H3D682D3A Synthonix Corporation

4-(Octylamino)pyridine

  • Cas Number : 64690-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57238.html

Safety Information

MSDS https://synthonix.com/57238.html
Availability details for
Quantity
  • Availability
  • Add to Cart