SY3H3D682D3C Synthonix Corporation

N-octylpyridinium bromide

  • MDL number No MDL found
  • Cas Number : 2534-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Product Link https://synthonix.com/57463.html

Safety Information

MSDS https://synthonix.com/57463.html
Availability details for
Quantity
  • Availability
  • Add to Cart