SY3H3D682D3D Synthonix Corporation

2-Oxo-2-(4-phenoxyphenyl)acetaldehyde hydrate

  • MDL number No MDL found
  • Cas Number : 92254-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57496.html

Safety Information

MSDS https://synthonix.com/57496.html
Availability details for
Quantity
  • Availability
  • Add to Cart