SY3H3D682D40 Synthonix Corporation

2-(4-Oxocyclohexyl)isoindoline-1,3-dione

  • MDL number MFCD00158659
  • Cas Number : 104618-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57622.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57622.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart