SY3H3D682D41 Synthonix Corporation

2-(4-Oxo-6-(piperazin-1-yl)quinazolin-3(4H)-yl)acetic acid

  • MDL number No MDL found
  • Cas Number : 889958-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57713.html

Safety Information

MSDS https://synthonix.com/57713.html
Availability details for
Quantity
  • Availability
  • Add to Cart