SY3H3D682D45 Synthonix Corporation

(4-(Octyloxy)phenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 121554-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57978.html

Safety Information

MSDS https://synthonix.com/57978.html
Availability details for
Quantity
  • Availability
  • Add to Cart