SY3H3D682D46 Synthonix Corporation

4,4'-Oxydibenzoyl chloride

  • MDL number No MDL found
  • Cas Number : 7158-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58040.html

Safety Information

MSDS https://synthonix.com/58040.html
Availability details for
Quantity
  • Availability
  • Add to Cart