SY3H3D682D4D Synthonix Corporation

2-(4-Octylphenyl)ethanol

  • MDL number No MDL found
  • Cas Number : 162358-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58843.html

Safety Information

MSDS https://synthonix.com/58843.html
Availability details for
Quantity
  • Availability
  • Add to Cart