SY3H3D682D53 Synthonix Corporation

Octyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 6278-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59470.html

Safety Information

MSDS https://synthonix.com/59470.html
Availability details for
Quantity
  • Availability
  • Add to Cart