SY3H3D682D54 Synthonix Corporation

2,2'-Oxybis(N,N-dibutylacetamide)

  • Cas Number : 82846-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59606.html

Safety Information

MSDS https://synthonix.com/59606.html
Availability details for
Quantity
  • Availability
  • Add to Cart