SY3H580D88F2 Synthonix Corporation

trans-tert-butyl 6-amino-1-azaspiro[3.3]heptane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1638769-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/59693.html

Safety Information

MSDS https://synthonix.com/59693.html
Availability details for
Quantity
  • Availability
  • Add to Cart