SY3H580D88F4 Synthonix Corporation

tert-butyl 3,3-difluoro-2-(hydroxymethyl)azetidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1638759-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59670.html

Safety Information

MSDS https://synthonix.com/59670.html
Availability details for
Quantity
  • Availability
  • Add to Cart