SY3H580D88F5 Synthonix Corporation

6-[(tert-butoxy)carbonyl]-2-oxa-6-azaspiro[3.3]heptane-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 2168618-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59673.html

Safety Information

MSDS https://synthonix.com/59673.html
Availability details for
Quantity
  • Availability
  • Add to Cart