SY3H580D88F6 Synthonix Corporation

tert-butyl 4,4-difluoro-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 2137836-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59735.html

Safety Information

MSDS https://synthonix.com/59735.html
Availability details for
Quantity
  • Availability
  • Add to Cart