SY3H580D88F7 Synthonix Corporation

tert-butyl 5-fluoro-2,7-diazaspiro[3.5]nonane-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 1250998-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59746.html

Safety Information

MSDS https://synthonix.com/59746.html
Availability details for
Quantity
  • Availability
  • Add to Cart