SY3H580D88F8 Synthonix Corporation

tert-butyl 3-amino-1-oxa-7-azaspiro[3.5]nonane-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 1638759-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59797.html

Safety Information

MSDS https://synthonix.com/59797.html
Availability details for
Quantity
  • Availability
  • Add to Cart