SY3H580D88FA Synthonix Corporation

6'-bromo-3',4'-dihydro-1'H-spiro[azetidine-2,2'-naphthalene]

  • MDL number No MDL found
  • Cas Number : 1478735-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59895.html

Safety Information

MSDS https://synthonix.com/59895.html
Availability details for
Quantity
  • Availability
  • Add to Cart