SY3H580D88FB Synthonix Corporation

3-benzyl-3,9-diazaspiro[5.5]undecan-11-ol dihydrochloride

  • MDL number No MDL found
  • Cas Number : 2250243-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59899.html

Safety Information

MSDS https://synthonix.com/59899.html
Availability details for
Quantity
  • Availability
  • Add to Cart