SY3H580D88FC Synthonix Corporation

tert-butyl 6-oxo-1-azaspiro[4.4]nonane-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 132564-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59944.html

Safety Information

MSDS https://synthonix.com/59944.html
Availability details for
Quantity
  • Availability
  • Add to Cart