SY3H580D88FF Synthonix Corporation

6-chloro-1,2-dihydrospiro[indole-3,3'-pyrrolidin]-2-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 2173991-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59874.html

Safety Information

MSDS https://synthonix.com/59874.html
Availability details for
Quantity
  • Availability
  • Add to Cart