SY3H580D8901 Synthonix Corporation

ethyl 8-[(tert-butyldimethylsilyl)oxy]-2-azaspiro[4.5]decane-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 1272655-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59937.html

Safety Information

MSDS https://synthonix.com/59937.html
Availability details for
Quantity
  • Availability
  • Add to Cart