SY3H580D8903 Synthonix Corporation

methyl 4,4-difluoropyrrolidine-3-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1638764-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60022.html

Safety Information

MSDS https://synthonix.com/60022.html
Availability details for
Quantity
  • Availability
  • Add to Cart