SY3H580D8904 Synthonix Corporation

spiro[2.3]hexane-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1782417-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59639.html

Safety Information

MSDS https://synthonix.com/59639.html
Availability details for
Quantity
  • Availability
  • Add to Cart