SY3H580D8905 Synthonix Corporation

spiro[3.3]heptan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1986337-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59785.html

Safety Information

MSDS https://synthonix.com/59785.html
Availability details for
Quantity
  • Availability
  • Add to Cart