SY3H580D8907 Synthonix Corporation

(8-?syn)?-3-?oxo-?Bicyclo[3.2.1]?octane-?8-?carboxylic acid ethyl ester

  • MDL number No MDL found
  • Cas Number : 1992829-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/60127.html

Safety Information

MSDS https://synthonix.com/60127.html
Availability details for
Quantity
  • Availability
  • Add to Cart