SY3H580D8928 Synthonix Corporation

[(2R,3S,4R,5R)-5-(4-benzamido-2-oxo-1,2-dihydropyrimidin-1-yl)-3-(benzoyloxy)-4-fluoro-4-methyloxolan-2-yl]methyl benzoate

  • MDL number MFCD31620826
  • Cas Number : 2173637-26-6
  • Molecular Weight : 571.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59629.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59629.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart