SY3H580D8930 Synthonix Corporation

tert-butyl 3,3-difluoro-1,6-diazaspiro[3.4]octane-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1823273-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59641.html

Safety Information

MSDS https://synthonix.com/59641.html
Availability details for
Quantity
  • Availability
  • Add to Cart