SY3H580D8949 Synthonix Corporation

(2R,4S)-Rel-1-(tert-butoxycarbonyl)-2-methylpiperidine-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1250957-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/59695.html

Safety Information

MSDS https://synthonix.com/59695.html
Availability details for
Quantity
  • Availability
  • Add to Cart