SY3H580D894C Synthonix Corporation

tert-butyl 4-(2-amino-1-fluoroethyl)piperidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1781380-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59699.html

Safety Information

MSDS https://synthonix.com/59699.html
Availability details for
Quantity
  • Availability
  • Add to Cart