SY3H580D8953 Synthonix Corporation

tert-butyl 3-hydroxy-1-oxa-9-azaspiro[5.5]undecane-9-carboxylate

  • MDL number No MDL found
  • Cas Number : 374795-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59726.html

Safety Information

MSDS https://synthonix.com/59726.html
Availability details for
Quantity
  • Availability
  • Add to Cart