SY3H580D8955 Synthonix Corporation

tert-butyl 6-oxa-2,9-diazaspiro[4.5]decane-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 1366647-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59730.html

Safety Information

MSDS https://synthonix.com/59730.html
Availability details for
Quantity
  • Availability
  • Add to Cart