SY3H580D8956 Synthonix Corporation

tert-butyl 3-amino-4,4-dimethylpiperidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1498964-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59731.html

Safety Information

MSDS https://synthonix.com/59731.html
Availability details for
Quantity
  • Availability
  • Add to Cart