SY3H580D8966 Synthonix Corporation

tert-butyl 3,3-difluoro-5-hydroxypiperidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1258638-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59756.html

Safety Information

MSDS https://synthonix.com/59756.html
Availability details for
Quantity
  • Availability
  • Add to Cart