SY3H580D896F Synthonix Corporation

tert-butyl (3S,4S)-3-fluoro-4-(hydroxymethyl)piperidine-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 1610418-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59779.html

Safety Information

MSDS https://synthonix.com/59779.html
Availability details for
Quantity
  • Availability
  • Add to Cart