SY3H580D898A Synthonix Corporation

2-{bicyclo[1.1.1]pentan-1-yl}propanoic acid

  • MDL number No MDL found
  • Cas Number : 2102409-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin China
Product Link https://synthonix.com/59831.html

Safety Information

MSDS https://synthonix.com/59831.html
Availability details for
Quantity
  • Availability
  • Add to Cart